Assay Details
| Gene Symbol: slc26a6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114417.1 | NP_001107889.1 | 10 - 11 | 1383 | 125 | |
| GenBank mRNA | BC155340.1 | - | 10 - 11 | 1383 | 125 | |
| FJ170818.1 | - | 10 - 11 | 1382 | 125 | ||
More Information
Set Membership: |
|

slc26a6l
Genomic Map