Assay Details
| Gene Symbol: hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114419.1 | NP_001107891.1 | 9 - 10 | 994 | 66 | |
| GenBank mRNA | BC155789.1 | - | 9 - 10 | 994 | 66 | |
More Information
Set Membership: |
|

hemk1
Genomic Map