Assay Details
| Gene Symbol: cip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114421.1 | NP_001107893.1 | 12 - 13 | 1847 | 68 | |
| XM_005162708.4 | XP_005162765.1 | 12 - 13 | 1896 | 68 | ||
| XM_021470191.1 | XP_021325866.1 | 13 - 14 | 1586 | 68 | ||
| GenBank mRNA | BC155117.1 | - | 12 - 13 | 1847 | 68 | |
More Information
Set Membership: |
|

cip2a
Genomic Map