Assay Details
| Gene Symbol: cip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114421.1 | NP_001107893.1 | 4 - 5 | 773 | 84 | |
| XM_005162708.4 | XP_005162765.1 | 4 - 5 | 822 | 84 | ||
| XM_021470191.1 | XP_021325866.1 | 5 - 6 | 512 | 84 | ||
| GenBank mRNA | BC155117.1 | - | 4 - 5 | 773 | 84 | |
More Information
Set Membership: |
|

cip2a
Genomic Map