Assay Details
| Gene Symbol: ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114432.1 | NP_001107904.1 | 2 - 3 | 145 | 67 | |
| GenBank mRNA | BC154812.1 | - | 2 - 3 | 145 | 67 | |
| KM655677.1 | - | 2 - 3 | 117 | 67 | ||
More Information
Set Membership: |
|

ramp2
Genomic Map