Assay Details
| Gene Symbol: oxr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114444.1 | NP_001107916.1 | 8 - 9 | 1291 | 74 | |
| XM_021466734.1 | XP_021322409.1 | 9 - 10 | 1701 | 74 | ||
| XM_021466735.1 | XP_021322410.1 | 10 - 11 | 1739 | 74 | ||
| XM_021466736.1 | XP_021322411.1 | 9 - 10 | 1702 | 74 | ||
| XM_021466737.1 | XP_021322412.1 | 8 - 9 | 1637 | 74 | ||
| GenBank mRNA | BC155345.1 | - | 8 - 9 | 1291 | 74 | |
| JQ649325.1 | - | 8 - 9 | 1469 | 74 | ||
| JQ649326.1 | - | 9 - 10 | 1579 | 74 | ||
More Information
Set Membership: |
|

oxr1a
Genomic Map