Assay Details
| Gene Symbol: oxr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114444.1 | NP_001107916.1 | 9 - 10 | 1462 | 71 | |
| XM_021466734.1 | XP_021322409.1 | 10 - 11 | 1872 | 71 | ||
| XM_021466735.1 | XP_021322410.1 | 11 - 12 | 1910 | 71 | ||
| XM_021466737.1 | XP_021322412.1 | 9 - 10 | 1808 | 71 | ||
| GenBank mRNA | BC155345.1 | - | 9 - 10 | 1462 | 71 | |
| JQ649325.1 | - | 9 - 10 | 1640 | 71 | ||
| JQ649326.1 | - | 10 - 11 | 1750 | 71 | ||
More Information
Set Membership: |
|

oxr1a
Genomic Map