Assay Details
| Gene Symbol: tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114446.1 | NP_001107918.2 | 8 - 9 | 884 | 78 | |
| XM_009293772.3 | XP_009292047.1 | 6 - 7 | 887 | 78 | ||
| XM_021467700.1 | XP_021323375.1 | 7 - 8 | 1099 | 78 | ||
| GenBank mRNA | BC155063.1 | - | 8 - 9 | 884 | 78 | |
More Information
Set Membership: |
|

tbrg1
Genomic Map