Assay Details
| Gene Symbol: parp14rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114447.2 | NP_001107919.2 | 15 - 16 | 4971 | 112 | |
| GenBank mRNA | BC155666.1 | - | 10 - 11 | 2259 | 112 | |
More Information
Set Membership: |
|

parp14rs1
Genomic Map