Assay Details
| Gene Symbol: parp14rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114447.2 | NP_001107919.2 | 6 - 7 | 3040 | 150 | |
| GenBank mRNA | BC155666.1 | - | 1 - 2 | 328 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC155666.1 transcripts.
Set Membership: |
|

parp14rs1
Genomic Map