Assay Details
| Gene Symbol: c1s.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114449.2 | NP_001107921.2 | 2 - 3 | 296 | 74 | |
| GenBank mRNA | BC155074.1 | - | 2 - 3 | 276 | 74 | |
More Information
Set Membership: |
|

c1s.2
Genomic Map