Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 11 - 12 | 807 | 138 | |
| XM_021480173.1 | XP_021335848.1 | 11 - 12 | 903 | 138 | ||
| XM_021480174.1 | XP_021335849.1 | 10 - 11 | 848 | 138 | ||
| XM_021480175.1 | XP_021335850.1 | 10 - 11 | 847 | 138 | ||
| XM_021480176.1 | XP_021335851.1 | 8 - 9 | 737 | 138 | ||
| GenBank mRNA | BC155327.1 | - | 11 - 12 | 776 | 137 | |
| EH494017.1 | - | 11 - 12 | 836 | 138 | ||
More Information
Set Membership: |
|

muc13b
Genomic Map