Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 12 - 13 | 859 | 107 | |
| XM_021480173.1 | XP_021335848.1 | 12 - 13 | 955 | 107 | ||
| XM_021480174.1 | XP_021335849.1 | 11 - 12 | 900 | 107 | ||
| XM_021480175.1 | XP_021335850.1 | 11 - 12 | 899 | 107 | ||
| XM_021480176.1 | XP_021335851.1 | 9 - 10 | 789 | 107 | ||
| GenBank mRNA | BC155327.1 | - | 12 - 13 | 827 | 106 | |
More Information
Set Membership: |
|

muc13b
Genomic Map