Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 16 - 17 | 1190 | 65 | |
| XM_021480173.1 | XP_021335848.1 | 16 - 17 | 1286 | 65 | ||
| XM_021480174.1 | XP_021335849.1 | 15 - 16 | 1231 | 65 | ||
| XM_021480175.1 | XP_021335850.1 | 15 - 16 | 1230 | 65 | ||
| XM_021480176.1 | XP_021335851.1 | 13 - 14 | 1120 | 65 | ||
| GenBank mRNA | BC155327.1 | - | 16 - 17 | 1158 | 65 | |
More Information
Set Membership: |
|

muc13b
Genomic Map