Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 18 - 19 | 1472 | 62 | |
| XM_021480173.1 | XP_021335848.1 | 18 - 19 | 1568 | 62 | ||
| XM_021480174.1 | XP_021335849.1 | 17 - 18 | 1513 | 62 | ||
| XM_021480175.1 | XP_021335850.1 | 17 - 18 | 1512 | 62 | ||
| XM_021480176.1 | XP_021335851.1 | 15 - 16 | 1402 | 62 | ||
| GenBank mRNA | BC155327.1 | - | 18 - 19 | 1440 | 62 | |
| EH520344.1 | - | 1 - 2 | 710 | 62 | ||
More Information
Set Membership: |
|

muc13b
Genomic Map