Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 3 - 4 | 274 | 81 | |
| XM_021480173.1 | XP_021335848.1 | 3 - 4 | 373 | 81 | ||
| XM_021480174.1 | XP_021335849.1 | 3 - 4 | 372 | 81 | ||
| XM_021480175.1 | XP_021335850.1 | 3 - 4 | 374 | 81 | ||
| GenBank mRNA | BC155327.1 | - | 3 - 4 | 243 | 81 | |
| EH494017.1 | - | 3 - 4 | 303 | 81 | ||
More Information
Set Membership: |
|

muc13b
Genomic Map