Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 5 - 6 | 386 | 96 | |
| GenBank mRNA | BC155327.1 | - | 5 - 6 | 355 | 96 | |
| EH494017.1 | - | 5 - 6 | 415 | 96 | ||
More Information
Set Membership: |
|

muc13b
Genomic Map