Assay Details
| Gene Symbol: muc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308550.1 | NP_001295479.1 | 7 - 8 | 488 | 83 | |
| XM_021480174.1 | XP_021335849.1 | 6 - 7 | 529 | 83 | ||
| XM_021480176.1 | XP_021335851.1 | 4 - 5 | 418 | 83 | ||
| GenBank mRNA | BC155327.1 | - | 7 - 8 | 457 | 83 | |
| EH494017.1 | - | 7 - 8 | 517 | 83 | ||
More Information
Set Membership: |
|

muc13b
Genomic Map