Assay Details
| Gene Symbol: msl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114468.1 | NP_001107940.1 | 4 - 5 | 1178 | 77 | |
| XM_021468803.1 | XP_021324478.1 | 3 - 4 | 480 | 77 | ||
| GenBank mRNA | BC155827.1 | - | 4 - 5 | 1178 | 77 | |
More Information
Set Membership: |
|

msl1a
Genomic Map