Assay Details
| Gene Symbol: tle3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313694.1 | NP_001300623.1 | 13 - 14 | 1368 | 101 | |
| NM_131012.2 | NP_571087.2 | 13 - 14 | 1365 | 101 | ||
| XM_005159039.4 | XP_005159096.1 | 14 - 15 | 1753 | 101 | ||
| XM_005159041.4 | XP_005159098.1 | 14 - 15 | 1750 | 101 | ||
| XM_017351898.2 | XP_017207387.1 | 14 - 15 | 1750 | 101 | ||
| GenBank mRNA | U96340.1 | - | 13 - 14 | 101 | ||
| Y12466.1 | - | 101 | ||||
More Information
Set Membership: |
|

tle3a
Genomic Map