Assay Details
| Gene Symbol: tle3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313694.1 | NP_001300623.1 | 14 - 15 | 1443 | 79 | |
| NM_131012.2 | NP_571087.2 | 14 - 15 | 1440 | 79 | ||
| XM_005159039.4 | XP_005159096.1 | 15 - 16 | 1828 | 79 | ||
| XM_005159041.4 | XP_005159098.1 | 15 - 16 | 1825 | 79 | ||
| XM_017351898.2 | XP_017207387.1 | 15 - 16 | 1825 | 79 | ||
| GenBank mRNA | BC162533.1 | - | 14 - 15 | 1341 | 79 | |
| U96340.1 | - | 14 - 15 | 1343 | 79 | ||
| Y12466.1 | - | 1343 | 79 | |||
More Information
Set Membership: |
|

tle3a
Genomic Map