Assay Details
| Gene Symbol: tle3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313694.1 | NP_001300623.1 | 19 - 20 | 2316 | 134 | |
| NM_131012.2 | NP_571087.2 | 19 - 20 | 2313 | 134 | ||
| XM_005159039.4 | XP_005159096.1 | 20 - 21 | 2701 | 134 | ||
| XM_005159041.4 | XP_005159098.1 | 20 - 21 | 2698 | 134 | ||
| XM_017351898.2 | XP_017207387.1 | 20 - 21 | 2698 | 134 | ||
| GenBank mRNA | BC162533.1 | - | 19 - 20 | 2214 | 134 | |
| U96340.1 | - | 19 - 20 | 134 | |||
| Y12466.1 | - | 134 | ||||
More Information
Set Membership: |
|

tle3a
Genomic Map