Assay Details
| Gene Symbol: tle3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313694.1 | NP_001300623.1 | 9 - 10 | 861 | 81 | |
| NM_131012.2 | NP_571087.2 | 9 - 10 | 858 | 81 | ||
| XM_005159039.4 | XP_005159096.1 | 10 - 11 | 1246 | 81 | ||
| XM_005159041.4 | XP_005159098.1 | 10 - 11 | 1243 | 81 | ||
| XM_017351898.2 | XP_017207387.1 | 10 - 11 | 1243 | 81 | ||
| GenBank mRNA | BC162533.1 | - | 9 - 10 | 759 | 81 | |
| U96340.1 | - | 9 - 10 | 761 | 81 | ||
| Y12466.1 | - | 761 | 81 | |||
More Information
Set Membership: |
|

tle3a
Genomic Map