Assay Details
| Gene Symbol: ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131025.4 | NP_571100.1 | 3 - 4 | 852 | 60 | |
| GenBank mRNA | AF365874.1 | - | 3 - 4 | 790 | 60 | |
| BC075743.1 | - | 3 - 4 | 809 | 60 | ||
| BC152628.1 | - | 3 - 4 | 808 | 60 | ||
| CT624760.2 | - | 2 - 3 | 326 | 60 | ||
| EH579652.1 | - | 3 - 4 | 842 | 60 | ||
| X87581.1 | - | 770 | 60 | |||
More Information
Set Membership: |
|

ccnd1
Genomic Map