Assay Details
| Gene Symbol: ahr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131028.2 | NP_571103.2 | 5 - 6 | 894 | 64 | |
| XM_021466612.1 | XP_021322287.1 | 5 - 6 | 911 | 64 | ||
| GenBank mRNA | AF258854.1 | - | 5 - 6 | 894 | 64 | |
| Y08433.1 | - | 151 | 64 | |||
More Information
Set Membership: |
|

ahr1a
Genomic Map