Assay Details
| Gene Symbol: ahr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131028.2 | NP_571103.2 | 7 - 8 | 1228 | 129 | |
| XM_021466612.1 | XP_021322287.1 | 7 - 8 | 1245 | 129 | ||
| GenBank mRNA | AF258854.1 | - | 7 - 8 | 1228 | 129 | |
| Y08433.1 | - | 485 | 129 | |||
More Information
Set Membership: |
|

ahr1a
Genomic Map