Assay Details
| Gene Symbol: neurog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131041.1 | NP_571116.1 | 1 - 2 | 94 | 107 | |
| GenBank mRNA | AF024535.2 | - | 1 - 2 | 100 | 107 | |
| AF036149.1 | - | 1 - 2 | 94 | 107 | ||
| BC056575.1 | - | 1 - 2 | 105 | 107 | ||
| BC066427.1 | - | 1 - 2 | 104 | 107 | ||
| U94588.1 | - | 1 - 2 | 103 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC056575.1, BC066427.1, NM_131041.1 transcripts.
Set Membership: |
|

neurog1
Genomic Map