Assay Details
| Gene Symbol: shha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131063.3 | NP_571138.1 | 1 - 2 | 622 | 134 | |
| GenBank mRNA | BC162395.1 | - | 1 - 2 | 334 | 134 | |
| KM655587.1 | - | 1 - 2 | 303 | 134 | ||
| L27585.1 | - | 1 - 2 | 622 | 134 | ||
| U30711.1 | - | 1 - 2 | 303 | 134 | ||
| Z35669.1 | - | 549 | 134 | |||
More Information
Set Membership: |
|

shha
Genomic Map