Assay Details
| Gene Symbol: hoxb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131142.2 | NP_571217.2 | 1 - 2 | 965 | 84 | |
| XM_005156266.4 | XP_005156323.1 | 3 - 4 | 3145 | 84 | ||
| XM_005156267.4 | XP_005156324.1 | 3 - 4 | 2934 | 84 | ||
| XM_017358429.2 | XP_017213918.1 | 2 - 3 | 3139 | 84 | ||
| XM_021480022.1 | XP_021335697.1 | 4 - 5 | 3116 | 84 | ||
| XM_021480023.1 | XP_021335698.1 | 4 - 5 | 3327 | 84 | ||
| XM_021480024.1 | XP_021335699.1 | 2 - 3 | 2856 | 84 | ||
| GenBank mRNA | AJ306433.1 | - | 1 - 2 | 591 | 84 | |
| BC133876.1 | - | 1 - 2 | 965 | 84 | ||
More Information
Set Membership: |
|

hoxb1b
Genomic Map