Assay Details
| Gene Symbol: opn1sw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131192.1 | NP_571267.1 | 1 - 2 | 506 | 63 | |
| GenBank mRNA | AF109372.1 | - | 1 - 2 | 506 | 63 | |
| BC059418.1 | - | 1 - 2 | 497 | 63 | ||
| BC062277.1 | - | 1 - 2 | 496 | 63 | ||
| BC153615.1 | - | 1 - 2 | 393 | 63 | ||
| KT008398.1 | - | 1 - 2 | 379 | 63 | ||
More Information
Set Membership: |
|

opn1sw2
Genomic Map