Assay Details
| Gene Symbol: myb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309822.1 | NP_001296751.1 | 15 - 16 | 1985 | 59 | |
| NM_131266.2 | NP_571341.2 | 14 - 15 | 1832 | 59 | ||
| GenBank mRNA | AF191559.1 | - | 14 - 15 | 1763 | 59 | |
| BC059803.1 | - | 15 - 16 | 1840 | 59 | ||
More Information
Set Membership: |
|

myb
Genomic Map