Assay Details
| Gene Symbol: pax6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131304.1 | NP_571379.1 | 5 - 6 | 730 | 101 | |
| XM_009297878.3 | XP_009296153.1 | 6 - 7 | 789 | 101 | ||
| XM_009297882.3 | XP_009296157.1 | 7 - 8 | 861 | 101 | ||
| XM_009297884.3 | XP_009296159.1 | 6 - 7 | 837 | 101 | ||
| XM_009297887.3 | XP_009296162.1 | 2 - 3 | 193 | 101 | ||
| XM_009297888.3 | XP_009296163.1 | 3 - 4 | 754 | 101 | ||
| XM_017354048.2 | XP_017209537.1 | 4 - 5 | 529 | 101 | ||
| XM_021470446.1 | XP_021326121.1 | 6 - 7 | 419 | 101 | ||
| GenBank mRNA | X63183.1 | - | 730 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009297887.3, XM_009297888.3 transcripts.
Set Membership: |
|

pax6a
Genomic Map