Assay Details
| Gene Symbol: pax6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131304.1 | NP_571379.1 | 6 - 7 | 945 | 84 | |
| XM_009297878.3 | XP_009296153.1 | 7 - 8 | 1004 | 84 | ||
| XM_009297882.3 | XP_009296157.1 | 8 - 9 | 1076 | 84 | ||
| XM_009297883.3 | XP_009296158.1 | 5 - 6 | 937 | 84 | ||
| XM_009297884.3 | XP_009296159.1 | 7 - 8 | 1052 | 84 | ||
| XM_009297885.3 | XP_009296160.1 | 6 - 7 | 1009 | 84 | ||
| XM_009297886.3 | XP_009296161.1 | 6 - 7 | 746 | 84 | ||
| XM_009297887.3 | XP_009296162.1 | 3 - 4 | 408 | 84 | ||
| XM_009297888.3 | XP_009296163.1 | 4 - 5 | 969 | 84 | ||
| XM_009297889.3 | XP_009296164.1 | 3 - 4 | 927 | 84 | ||
| XM_017354048.2 | XP_017209537.1 | 5 - 6 | 744 | 84 | ||
| XM_017354050.2 | XP_017209539.1 | 6 - 7 | 961 | 84 | ||
| XM_021470446.1 | XP_021326121.1 | 7 - 8 | 634 | 84 | ||
| XM_021470447.1 | XP_021326122.1 | 5 - 6 | 585 | 84 | ||
| XM_021470448.1 | XP_021326123.1 | 6 - 7 | 712 | 84 | ||
| GenBank mRNA | BC066722.1 | - | 5 - 6 | 868 | 84 | |
| X61389.1 | - | 880 | 84 | |||
| X63183.1 | - | 945 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009297886.3, XM_009297887.3, XM_009297888.3, XM_009297889.3 transcripts.
Set Membership: |
|

pax6a
Genomic Map