Assay Details
| Gene Symbol: pax6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131304.1 | NP_571379.1 | 7 - 8 | 1109 | 125 | |
| XM_009297878.3 | XP_009296153.1 | 8 - 9 | 1168 | 125 | ||
| XM_009297882.3 | XP_009296157.1 | 9 - 10 | 1240 | 125 | ||
| XM_009297883.3 | XP_009296158.1 | 6 - 7 | 1101 | 125 | ||
| XM_009297884.3 | XP_009296159.1 | 8 - 9 | 1216 | 125 | ||
| XM_009297885.3 | XP_009296160.1 | 7 - 8 | 1173 | 125 | ||
| XM_009297886.3 | XP_009296161.1 | 7 - 8 | 910 | 125 | ||
| XM_009297887.3 | XP_009296162.1 | 4 - 5 | 572 | 125 | ||
| XM_009297888.3 | XP_009296163.1 | 5 - 6 | 1133 | 125 | ||
| XM_009297889.3 | XP_009296164.1 | 4 - 5 | 1091 | 125 | ||
| XM_017354048.2 | XP_017209537.1 | 6 - 7 | 908 | 125 | ||
| XM_017354050.2 | XP_017209539.1 | 7 - 8 | 1125 | 125 | ||
| XM_021470446.1 | XP_021326121.1 | 8 - 9 | 798 | 125 | ||
| XM_021470447.1 | XP_021326122.1 | 6 - 7 | 749 | 125 | ||
| XM_021470448.1 | XP_021326123.1 | 7 - 8 | 876 | 125 | ||
| GenBank mRNA | X61389.1 | - | 1044 | 125 | ||
| X63183.1 | - | 1109 | 125 | |||
More Information
Set Membership: |
|

pax6a
Genomic Map