Assay Details
| Gene Symbol: cd74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131372.2 | NP_571447.1 | 3 - 4 | 349 | 71 | |
| GenBank mRNA | AF116539.1 | - | 3 - 4 | 361 | 71 | |
| BC092708.1 | - | 3 - 4 | 381 | 71 | ||
| BC110099.1 | - | 3 - 4 | 393 | 71 | ||
| BC124092.1 | - | 3 - 4 | 384 | 71 | ||
| BC127569.1 | - | 3 - 4 | 349 | 71 | ||
| BC129254.1 | - | 3 - 4 | 383 | 71 | ||
More Information
Set Membership: |
|

cd74b
Genomic Map