Assay Details
| Gene Symbol: l1camb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131383.1 | NP_571458.1 | 16 - 17 | 2128 | 98 | |
| XM_009296546.3 | XP_009294821.1 | 12 - 13 | 1791 | 98 | ||
| GenBank mRNA | AY376855.1 | - | 16 - 17 | 2128 | 98 | |
| CU458881.1 | - | 15 - 16 | 1939 | 98 | ||
| X89204.1 | - | 1729 | 98 | |||
More Information
Set Membership: |
|

l1camb
Genomic Map