Assay Details
| Gene Symbol: psmb9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131391.2 | NP_571466.1 | 1 - 2 | 316 | 83 | |
| GenBank mRNA | BC078384.1 | - | 1 - 2 | 316 | 83 | |
| BC165916.1 | - | 1 - 2 | 96 | 83 | ||
More Information
Set Membership: |
|

psmb9a
Genomic Map