Assay Details
| Gene Symbol: psmb9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131391.2 | NP_571466.1 | 5 - 6 | 790 | 116 | |
| GenBank mRNA | AF155579.1 | - | 5 - 6 | 590 | 116 | |
| BC078384.1 | - | 5 - 6 | 790 | 116 | ||
More Information
Set Membership: |
|

psmb9a
Genomic Map