Assay Details
| Gene Symbol: aanat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131411.2 | NP_571486.1 | 2 - 3 | 425 | 62 | |
| GenBank mRNA | AF124756.1 | - | 2 - 3 | 445 | 62 | |
| BC059422.1 | - | 2 - 3 | 430 | 62 | ||
| BC071294.1 | - | 2 - 3 | 427 | 62 | ||
| EH115279.1 | - | 2 - 3 | 425 | 62 | ||
More Information
Set Membership: |
|

aanat2
Genomic Map