Assay Details
| Gene Symbol: kdrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131472.1 | NP_571547.1 | 19 - 20 | 2648 | 82 | |
| XM_009291142.3 | XP_009289417.1 | 19 - 20 | 3042 | 82 | ||
| GenBank mRNA | AF180354.1 | - | 16 - 17 | 2264 | 82 | |
| AF487829.1 | - | 19 - 20 | 3001 | 82 | ||
| AY056466.1 | - | 19 - 20 | 2648 | 82 | ||
| BC129158.1 | - | 19 - 20 | 3016 | 82 | ||
| CU458916.1 | - | 19 - 20 | 3016 | 82 | ||
| U89515.1 | - | 1 - 2 | 77 | 82 | ||
| Gene Symbol: LOC103909948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009298309.3 | XP_009296584.3 | 1 - 2 | 157 | 82 | |
More Information
Set Membership: |
|

kdrl
Genomic Map