Assay Details
| Gene Symbol: baxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131562.2 | NP_571637.1 | 3 - 4 | 258 | 69 | |
| GenBank mRNA | BC055592.1 | - | 3 - 4 | 258 | 69 | |
| BC164990.1 | - | 3 - 4 | 272 | 69 | ||
More Information
Set Membership: |
|

baxa
Genomic Map