Assay Details
| Gene Symbol: tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131563.2 | NP_571638.1 | 4 - 5 | 372 | 76 | |
| GenBank mRNA | BC064284.1 | - | 4 - 5 | 372 | 76 | |
| BC154312.1 | - | 4 - 5 | 386 | 76 | ||
More Information
Set Membership: |
|

tnnc2
Genomic Map