Assay Details
| Gene Symbol: slc3a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131601.2 | NP_571676.2 | 7 - 8 | 1159 | 88 | |
| GenBank mRNA | AF295374.2 | - | 7 - 8 | 1130 | 88 | |
| BC053236.1 | - | 7 - 8 | 1141 | 88 | ||
| BC159202.1 | - | 7 - 8 | 1142 | 88 | ||
More Information
Set Membership: |
|

slc3a2a
Genomic Map