Assay Details
| Gene Symbol: slc3a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131601.2 | NP_571676.2 | 8 - 9 | 1195 | 139 | |
| GenBank mRNA | AF295374.2 | - | 8 - 9 | 139 | ||
| BC053236.1 | - | 8 - 9 | 139 | |||
| BC159202.1 | - | 8 - 9 | 139 | |||
More Information
Set Membership: |
|

slc3a2a
Genomic Map