Assay Details
| Gene Symbol: angpt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131814.1 | NP_571889.1 | 2 - 3 | 442 | 114 | |
| GenBank mRNA | AF379603.1 | - | 2 - 3 | 442 | 114 | |
| BC162238.1 | - | 2 - 3 | 453 | 114 | ||
More Information
Set Membership: |
|

angpt2b
Genomic Map