Assay Details
| Gene Symbol: cnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126448.2 | NP_001119920.2 | 3 - 4 | 438 | 114 | |
| XM_009305008.3 | XP_009303283.1 | 3 - 4 | 386 | 114 | ||
| GenBank mRNA | AF322374.1 | - | 3 - 4 | 438 | 114 | |
| AI477790.1 | - | 3 - 4 | 256 | 114 | ||
| BC066426.1 | - | 3 - 4 | 272 | 114 | ||
| BQ826523.1 | - | 3 - 4 | 257 | 114 | ||
| EH585281.1 | - | 3 - 4 | 347 | 114 | ||
More Information
Set Membership: |
|

cnmd
Genomic Map