Assay Details
| Gene Symbol: cnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126448.2 | NP_001119920.2 | 5 - 6 | 660 | 89 | |
| XM_009305008.3 | XP_009303283.1 | 5 - 6 | 608 | 89 | ||
| GenBank mRNA | AF322374.1 | - | 5 - 6 | 660 | 89 | |
| BC066426.1 | - | 5 - 6 | 494 | 89 | ||
| EH585281.1 | - | 5 - 6 | 569 | 89 | ||
More Information
Set Membership: |
|

cnmd
Genomic Map