Assay Details
| Gene Symbol: xbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131874.1 | NP_571949.1 | 1 - 2 | 337 | 153 | |
| GenBank mRNA | AF399656.1 | - | 1 - 2 | 334 | 153 | |
| AF399918.1 | - | 1 - 2 | 337 | 153 | ||
| AF420255.1 | - | 1 - 2 | 320 | 153 | ||
| AF420256.1 | - | 1 - 2 | 318 | 153 | ||
| AY029577.1 | - | 1 - 2 | 337 | 153 | ||
| BC044133.1 | - | 1 - 2 | 350 | 153 | ||
| BC044134.1 | - | 1 - 2 | 350 | 153 | ||
| BC066493.1 | - | 1 - 2 | 316 | 153 | ||
| BC152195.1 | - | 1 - 2 | 350 | 153 | ||
| BC164779.1 | - | 1 - 2 | 259 | 153 | ||
More Information
Set Membership: |
|

xbp1
Genomic Map