Assay Details
| Gene Symbol: csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152951.1 | NP_694483.1 | 8 - 9 | 1359 | 77 | |
| XM_009291086.3 | XP_009289361.1 | 9 - 10 | 1471 | 77 | ||
| XM_009291087.3 | XP_009289362.1 | 9 - 10 | 1471 | 77 | ||
| XM_009291089.3 | XP_009289364.1 | 9 - 10 | 1471 | 77 | ||
| XM_009291090.3 | XP_009289365.1 | 8 - 9 | 1386 | 77 | ||
| GenBank mRNA | AF516328.1 | - | 9 - 10 | 1457 | 77 | |
| AF516329.1 | - | 9 - 10 | 1258 | 77 | ||
| AY099516.1 | - | 8 - 9 | 1359 | 77 | ||
| AY116143.1 | - | 8 - 9 | 1336 | 77 | ||
| BC081610.1 | - | 8 - 9 | 1344 | 77 | ||
| BC154280.1 | - | 9 - 10 | 1420 | 77 | ||
More Information
Set Membership: |
|

csnk1a1
Genomic Map