Assay Details
| Gene Symbol: h2az2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153644.1 | NP_705930.1 | 3 - 4 | 244 | 63 | |
| XM_005165036.3 | XP_005165093.1 | 3 - 4 | 341 | 63 | ||
| GenBank mRNA | AF414110.1 | - | 3 - 4 | 244 | 63 | |
| BC049019.1 | - | 3 - 4 | 272 | 63 | ||
| BC154791.1 | - | 3 - 4 | 272 | 63 | ||
| EH508860.1 | - | 3 - 4 | 339 | 63 | ||
More Information
Set Membership: |
|

h2az2a
Genomic Map